siRNA Details
| Gene Symbol: DOK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152721.5 | NP_689934.2 | 2, 3 | 381 | ||
| XM_011525875.2 | XP_011524177.1 | 2, 3 | 402 | |||
| GenBank mRNA | AK057795.1 | - | 1, 2 | 90 | ||
| AY599248.1 | - | 2, 3 | 379 | |||
| BC096744.1 | - | 2, 3 | 374 | 5260167 | ||

DOK6
Genomic Map