siRNA Details
| Gene Symbol: HEPACAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152722.4 | NP_689935.2 | 3 | 1099 | ||
| XM_005271449.2 | XP_005271506.1 | 3 | 1099 | |||
| XM_017017361.1 | XP_016872850.1 | 3 | 1099 | |||
| GenBank mRNA | AK098396.1 | - | 3 | 737 | ||
| AK122595.1 | - | 3 | 737 | |||
| AY047587.1 | - | 3 | 737 | |||
| BC104831.1 | - | 3 | 721 | 8143834 | ||
| BC113562.1 | - | 3 | 721 | 8322614 | ||

HEPACAM
Genomic Map