siRNA Details
| Gene Symbol: CPNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152727.5 | NP_689940.3 | 3 | 557 | ||
| GenBank mRNA | AF492484.1 | - | 3 | 508 | ||
| AK292269.1 | - | 2 | 466 | |||
| AK315394.1 | - | 3 | 460 | |||
| BC053658.1 | - | 4 | 631 | 5750192 | ||
| BG830959.1 | - | 3 | 503 | |||
| BG831872.1 | - | 3 | 505 | |||
| CR749617.1 | - | 2 | 218 | |||

CPNE2
Genomic Map