siRNA Details
Gene Symbol: ADGRG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304376.2 | NP_001291305.1 | 8 | 838 | ||
NM_001318481.1 | NP_001305410.1 | 8 | 838 | |||
NM_153837.3 | NP_722579.1 | 8 | 1310 | |||
XM_011522949.2 | XP_011521251.1 | 8 | 854 | |||
XM_011522950.2 | XP_011521252.1 | 8 | 880 | |||
GenBank mRNA | AK128298.1 | - | 8 | 989 | ||
AK160368.1 | - | 7 | 2268 | |||
AK172765.1 | - | 4 | 327 | |||
AK292783.1 | - | 8 | 830 | |||
AK303661.1 | - | 8 | 830 | |||
AY140956.1 | - | 7 | 735 | |||
AY358447.1 | - | 8 | 817 | |||
BC032401.1 | - | 8 | 1258 | 5210813 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
