siRNA Details
| Gene Symbol: JAZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175061.3 | NP_778231.2 | 4, 5 | 796 | ||
| XM_006715656.1 | XP_006715719.1 | 5, 6 | 672 | |||
| XM_017011831.1 | XP_016867320.1 | 4, 5 | 564 | |||
| XM_017011832.1 | XP_016867321.1 | 4, 5 | 496 | |||
| GenBank mRNA | AK000711.1 | - | 1, 2 | 23 | ||
| AK091311.1 | - | 4, 5 | 721 | |||
| AL832491.1 | - | 2, 3 | 374 | |||
| AL834234.1 | - | 3, 4 | 515 | |||
| AY372319.1 | - | 4, 5 | 574 | |||
| BC042441.1 | - | 5, 6 | 584 | 4831063 | ||
| BC047229.1 | - | 4, 5 | 583 | |||

JAZF1
Genomic Map