siRNA Details
| Gene Symbol: TMEM130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134450.1 | NP_001127922.1 | 8 | 1405 | ||
| NM_001134451.1 | NP_001127923.1 | 7 | 1063 | |||
| NM_152913.2 | NP_690877.1 | 8 | 1369 | |||
| GenBank mRNA | AB072903.1 | - | 3 | 219 | ||
| AK124634.1 | - | 8 | 2246 | |||
| AK225783.1 | - | 8 | 1348 | |||
| AK294658.1 | - | 8 | 1185 | |||
| AK295543.1 | - | 7 | 1027 | |||
| AK312364.1 | - | 8 | 1239 | |||
| AL832523.1 | - | 7 | 1103 | |||
| AL834348.1 | - | 8 | 1215 | |||
| AL834349.1 | - | 8 | 1221 | |||
| AY358583.1 | - | 9 | 1301 | |||
| BC030793.1 | - | 8 | 1338 | 5263130 | ||
| BC037895.1 | - | 7 | 975 | 5275566 | ||

TMEM130
Genomic Map