siRNA Details
| Gene Symbol: CNPY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152755.1 | NP_689968.1 | 5, 6 | 687 | ||
| GenBank mRNA | AK075537.1 | - | 5, 6 | 641 | ||
| AY359100.1 | - | 5, 6 | 691 | |||
| BC019903.1 | - | 5, 6 | 617 | 4991480 | ||
| BC032339.1 | - | 5, 6 | 687 | 5191897 | ||

CNPY4
Genomic Map