siRNA Details
| Gene Symbol: STAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286256.1 | NP_001273185.1 | 11 | 1214 | ||
| NM_001286257.1 | NP_001273186.1 | 9 | 848 | |||
| NM_145064.2 | NP_659501.1 | 12 | 1281 | |||
| NR_104422.1 | 8 | 781 | ||||
| XM_011538126.2 | XP_011536428.1 | 12 | 1369 | |||
| GenBank mRNA | AK057013.1 | - | 12 | 1274 | ||
| AK300688.1 | - | 9 | 778 | |||
| AK311513.1 | - | 8 | 774 | |||
| BC008069.2 | - | 11 | 1194 | 2961089 | ||
| BU738507.1 | - | 1 | 384 | |||

STAC3
Genomic Map