siRNA Details
| Gene Symbol: SPESP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145658.3 | NP_663633.1 | 2 | 1182 | ||
| GenBank mRNA | AF275321.1 | - | 2 | 1086 | ||
| AY358983.1 | - | 2 | 1165 | |||
| BC017998.1 | - | 2 | 1156 | 4252566 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPESP1| Gene Symbol: SPESP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145658.3 | NP_663633.1 | 2 | 1182 | ||
| GenBank mRNA | AF275321.1 | - | 2 | 1086 | ||
| AY358983.1 | - | 2 | 1165 | |||
| BC017998.1 | - | 2 | 1156 | 4252566 | ||