siRNA Details
Gene Symbol: EPHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173567.4 | NP_775838.3 | 6 | 925 | ||
GenBank mRNA | AK074822.1 | - | 6 | 885 | ||
BC041475.1 | - | 6 | 900 | 5244314 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: EPHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173567.4 | NP_775838.3 | 6 | 925 | ||
GenBank mRNA | AK074822.1 | - | 6 | 885 | ||
BC041475.1 | - | 6 | 900 | 5244314 |