siRNA Details
| Gene Symbol: MSRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031679.2 | NP_001026849.1 | 6 | 582 | ||
| NM_001193460.1 | NP_001180389.1 | 7 | 746 | |||
| NM_001193461.1 | NP_001180390.1 | 6 | 444 | |||
| NM_198080.3 | NP_932346.1 | 5 | 455 | |||
| GenBank mRNA | AK293084.1 | - | 6 | 552 | ||
| AK299065.1 | - | 5 | 394 | |||
| AK316365.1 | - | 5 | 426 | |||
| AL833622.1 | - | 6 | 500 | |||
| AY358229.1 | - | 5 | 390 | |||
| BC040053.1 | - | 7 | 647 | 6025598 | ||
| BX500552.1 | - | 6 | 446 | |||
| BX648776.1 | - | 6 | 501 | |||
| DC357690.1 | - | 5 | 455 | |||

MSRB3
Genomic Map