siRNA Details
| Gene Symbol: CNIH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182553.2 | NP_872359.1 | 5 | 623 | ||
| NR_073078.1 | 5 | 649 | ||||
| NR_073079.1 | 5 | 619 | ||||
| GenBank mRNA | AY726585.1 | - | 5 | 394 | ||
| BC047953.1 | - | 5 | 599 | |||
| BM547951.1 | - | 623 | ||||
| DR760677.1 | - | 5 | 595 | |||

CNIH2
Genomic Map