siRNA Details
| Gene Symbol: AKNAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152763.4 | NP_689976.2 | 2 | 1057 | ||
| NR_049760.1 | 3 | 1198 | ||||
| GenBank mRNA | AK095517.1 | - | 3 | 1184 | ||
| AK313772.1 | - | 2 | 1037 | |||
| AL832216.1 | - | 2 | 1047 | |||
| BC030647.2 | - | 2 | 1037 | |||
| BC150642.1 | - | 1 | 812 | 9057012 | ||

AKNAD1
Genomic Map