siRNA Details
| Gene Symbol: AKNAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152763.4 | NP_689976.2 | 4 | 1447 | ||
| NR_049760.1 | 5 | 1588 | ||||
| GenBank mRNA | AK095517.1 | - | 5 | 1574 | ||
| AK313772.1 | - | 4 | 1427 | |||
| AL832216.1 | - | 4 | 1437 | |||
| BC030647.2 | - | 4 | 1427 | |||
| BC150642.1 | - | 3 | 1202 | 9057012 | ||

AKNAD1
Genomic Map