siRNA Details
| Gene Symbol: C8orf46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152765.3 | NP_689978.2 | 5 | 813 | ||
| GenBank mRNA | AK055047.1 | - | 2 | 147 | ||
| AK096336.1 | - | 6 | 601 | |||
| AK294261.1 | - | 4 | 401 | |||
| AK308800.1 | - | 6 | 640 | |||
| AK315486.1 | - | 5 | 556 | |||
| BC028400.2 | - | 5 | 537 | 4816822 | ||

C8orf46
Genomic Map