siRNA Details
| Gene Symbol: MORN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198469.3 | NP_940871.2 | 2, 3 | 256 | ||
| XM_005251877.4 | XP_005251934.1 | 2, 3 | 252 | |||
| XM_005251878.3 | XP_005251935.1 | 2, 3 | 227 | |||
| XM_011518457.2 | XP_011516759.1 | 2, 3 | 252 | |||
| XM_011518458.1 | XP_011516760.1 | 3, 4 | 335 | |||
| GenBank mRNA | AI026684.1 | - | 3, 4 | 450 | ||
| AK128877.1 | - | 2, 3 | 253 | |||
| AK302081.1 | - | 2, 3 | 230 | |||
| BC133690.1 | - | 2, 3 | 247 | 40119769 | ||
| HY011325.1 | - | 2, 3 | 257 | |||

MORN5
Genomic Map