siRNA Details
| Gene Symbol: MORN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286828.1 | NP_001273757.1 | 3, 4 | 400 | ||
| NM_198469.3 | NP_940871.2 | 4, 5 | 512 | |||
| XM_005251878.3 | XP_005251935.1 | 4, 5 | 483 | |||
| XM_011518458.1 | XP_011516760.1 | 5, 6 | 591 | |||
| GenBank mRNA | AI026684.1 | - | 1, 2 | 194 | ||
| AI560944.1 | - | 1, 2 | 212 | |||
| AK128877.1 | - | 4, 5 | 509 | |||
| BC133690.1 | - | 4, 5 | 503 | 40119769 | ||
| BC143440.1 | - | 3, 4 | 360 | |||

MORN5
Genomic Map