siRNA Details
| Gene Symbol: C4orf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206997.1 | NP_001193926.1 | 1, 2 | 152 | ||
| NM_152770.2 | NP_689983.2 | 1, 2 | 152 | |||
| XM_011531816.2 | XP_011530118.2 | 2, 3 | 318 | |||
| XM_017007972.1 | XP_016863461.1 | 2, 3 | 304 | |||
| XM_017007973.1 | XP_016863462.1 | 2, 3 | 307 | |||
| XM_017007974.1 | XP_016863463.1 | 2, 3 | 312 | |||
| XM_017007976.1 | XP_016863465.1 | 2, 3 | 327 | |||
| XM_017007977.1 | XP_016863466.1 | 2, 3 | 318 | |||
| GenBank mRNA | AK128642.1 | - | 1, 2 | 104 | ||
| AY349359.1 | - | 1, 2 | 103 | |||
| BC034296.1 | - | 1, 2 | 152 | 5165558 | ||

C4orf22
Genomic Map