siRNA Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153260.2 | NP_694992.2 | 4 | 432 | ||
| XM_011521423.2 | XP_011519725.1 | 3 | 320 | |||
| XM_011521424.2 | XP_011519726.1 | 3 | 320 | |||
| GenBank mRNA | AK094131.1 | - | 4 | 426 | ||
| AK094891.1 | - | 3 | 666 | |||
| AK309861.1 | - | 3 | 997 | |||
| BC058935.1 | - | 4 | 388 | 5578024 | ||
| BX538324.1 | - | 3 | 578 | |||
| CR749223.1 | - | 4 | 401 | |||
| DA749946.1 | - | 4 | 432 | |||

LRRC57
Genomic Map