siRNA Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153260.2 | NP_694992.2 | 5 | 657 | ||
| XM_011521423.2 | XP_011519725.1 | 4 | 545 | |||
| XM_011521424.2 | XP_011519726.1 | 4 | 545 | |||
| GenBank mRNA | AK094131.1 | - | 5 | 651 | ||
| AK094891.1 | - | 4 | 891 | |||
| AK309861.1 | - | 4 | 1222 | |||
| BC058935.1 | - | 5 | 613 | 5578024 | ||
| BX538324.1 | - | 4 | 803 | |||
| CN350663.1 | - | 2 | 121 | |||
| CR749223.1 | - | 5 | 626 | |||

LRRC57
Genomic Map