siRNA Details
| Gene Symbol: TCP11L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152772.2 | NP_689985.1 | 7 | 1077 | ||
| XM_005268767.4 | XP_005268824.1 | 8 | 1227 | |||
| XM_005268768.4 | XP_005268825.1 | 7 | 1233 | |||
| XM_011538129.2 | XP_011536431.1 | 8 | 1995 | |||
| XM_011538130.2 | XP_011536432.1 | 8 | 1372 | |||
| XM_011538131.2 | XP_011536433.1 | 8 | 1230 | |||
| XM_011538132.2 | XP_011536434.1 | 7 | 1112 | |||
| XM_017019127.1 | XP_016874616.1 | 7 | 1906 | |||
| XM_017019128.1 | XP_016874617.1 | 9 | 2142 | |||
| XM_017019129.1 | XP_016874618.1 | 9 | 1630 | |||
| XM_017019130.1 | XP_016874619.1 | 8 | 1178 | |||
| XM_017019131.1 | XP_016874620.1 | 7 | 950 | |||
| XM_017019132.1 | XP_016874621.1 | 9 | 1369 | |||
| XM_017019133.1 | XP_016874622.1 | 6 | 897 | |||
| GenBank mRNA | AK314055.1 | - | 7 | 1017 | ||
| BC033617.1 | - | 7 | 1065 | 5248900 | ||

TCP11L2
Genomic Map