siRNA Details
| Gene Symbol: RNF144B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182757.3 | NP_877434.2 | 5, 6 | 864 | ||
| XM_005248986.3 | XP_005249043.1 | 4, 5 | 780 | |||
| GenBank mRNA | AB076367.1 | - | 5, 6 | 846 | ||
| AK096832.1 | - | 5, 6 | 864 | |||
| AK302935.1 | - | 2, 3 | 487 | |||
| AL832329.1 | - | 5, 6 | 689 | |||
| BC063311.1 | - | 5, 6 | 738 | 30331343 | ||

RNF144B
Genomic Map