siRNA Details
| Gene Symbol: TCTEX1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152773.4 | NP_689986.2 | 1, 2 | 263 | ||
| GenBank mRNA | BC015348.1 | - | 1, 2 | 199 | ||
| BC021177.2 | - | 1, 2 | 243 | 4830500 | ||
| BM548106.1 | - | 1, 2 | 288 | |||
| CK818899.1 | - | 1, 2 | 257 | |||

TCTEX1D2
Genomic Map