siRNA Details
| Gene Symbol: TCTEX1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152773.4 | NP_689986.2 | 5 | 547 | ||
| GenBank mRNA | BC021177.2 | - | 5 | 527 | 4830500 | |
| BM548106.1 | - | 6 | 690 | |||
| BM983853.1 | - | 145 | ||||
| CK818899.1 | - | 5 | 541 | |||

TCTEX1D2
Genomic Map