siRNA Details
| Gene Symbol: PDZD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173806.3 | NP_776167.2 | 3 | 663 | ||
| NR_033694.1 | 5 | 914 | ||||
| XM_006721029.3 | XP_006721092.1 | 4 | 867 | |||
| GenBank mRNA | BC021724.1 | - | 5 | 919 | 4829370 | |
| BC028605.1 | - | 5 | 919 | 5265642 | ||
| BC039562.1 | - | 5 | 911 | 5169242 | ||
| BX100380.1 | - | 1 | 184 | |||

PDZD9
Genomic Map