siRNA Details
| Gene Symbol: COL6A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278298.1 | NP_001265227.1 | 36 | 7024 | ||
| NM_153264.6 | NP_694996.5 | 36 | 7024 | |||
| NR_022012.2 | 36 | 7024 | ||||
| XM_011512621.2 | XP_011510923.1 | 35 | 6835 | |||
| GenBank mRNA | AK093199.1 | - | 2 | 245 | ||
| AK123718.1 | - | 4 | 835 | |||
| AM906082.1 | - | 4 | 1172 | |||
| AM906083.1 | - | 2 | 359 | |||
| EU085556.1 | - | 36 | 7024 | |||

COL6A5
Genomic Map