siRNA Details
| Gene Symbol: GLIPR1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304964.1 | NP_001291893.1 | 2, 3 | 517 | ||
| NM_152779.3 | NP_689992.1 | 2, 3 | 517 | |||
| XM_011538134.2 | XP_011536436.2 | 2, 3 | 3312 | |||
| GenBank mRNA | AY358731.1 | - | 2, 3 | 517 | ||
| BC014603.1 | - | 2, 3 | 478 | 4822995 | ||

GLIPR1L1
Genomic Map