siRNA Details
| Gene Symbol: KLHL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153270.2 | NP_695002.1 | 1 | 612 | ||
| GenBank mRNA | AK092279.1 | - | 1 | 555 | ||
| BC109031.2 | - | 1 | 524 | 40027951 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLHL34| Gene Symbol: KLHL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153270.2 | NP_695002.1 | 1 | 612 | ||
| GenBank mRNA | AK092279.1 | - | 1 | 555 | ||
| BC109031.2 | - | 1 | 524 | 40027951 | ||