siRNA Details
| Gene Symbol: TAS2R41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176883.2 | NP_795364.2 | 1 | 68 | ||
| GenBank mRNA | BC101157.3 | - | 1 | 68 | 40015710 | |
| BC101158.2 | - | 1 | 68 | 40015713 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS2R41| Gene Symbol: TAS2R41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176883.2 | NP_795364.2 | 1 | 68 | ||
| GenBank mRNA | BC101157.3 | - | 1 | 68 | 40015710 | |
| BC101158.2 | - | 1 | 68 | 40015713 | ||