siRNA Details
| Gene Symbol: TAS2R43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101155.1 | - | 2 | 77 | 40015679 | |
| Gene Symbol: TAS2R45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176886.2 | NP_795367.2 | 2 | 77 | ||
| GenBank mRNA | BC101154.1 | - | 2 | 77 | 40015678 | |
| BC101156.1 | - | 2 | 77 | 40015680 | ||

TAS2R43
Genomic Map