siRNA Details
| Gene Symbol: TAS2R19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176888.2 | NP_795369.1 | 1 | 318 | ||
| GenBank mRNA | BC101804.1 | - | 1 | 318 | 8069310 | |
| BC111998.1 | - | 1 | 330 | 8327466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS2R19| Gene Symbol: TAS2R19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176888.2 | NP_795369.1 | 1 | 318 | ||
| GenBank mRNA | BC101804.1 | - | 1 | 318 | 8069310 | |
| BC111998.1 | - | 1 | 330 | 8327466 | ||