siRNA Details
Gene Symbol: TAS2R50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176890.2 | NP_795371.2 | 1 | 434 | ||
GenBank mRNA | BC112100.1 | - | 1 | 434 | 8327568 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TAS2R50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176890.2 | NP_795371.2 | 1 | 434 | ||
GenBank mRNA | BC112100.1 | - | 1 | 434 | 8327568 |