siRNA Details
| Gene Symbol: MDGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153487.3 | NP_705691.1 | 16 | 3940 | ||
| XM_006715056.3 | XP_006715119.1 | 16 | 2884 | |||
| XM_017010734.1 | XP_016866223.1 | 16 | 2884 | |||
| GenBank mRNA | AF478693.1 | - | 16 | 3940 | ||
| AK090677.1 | - | 7 | 740 | |||
| AK126965.1 | - | 2 | 1382 | |||
| BX647180.1 | - | 4 | 519 | |||
| BX647253.1 | - | 4 | 519 | |||

MDGA1
Genomic Map