siRNA Details
| Gene Symbol: NAP1L5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153757.3 | NP_715638.1 | 1 | 835 | ||
| GenBank mRNA | AK054689.1 | - | 1 | 429 | ||
| AL834329.1 | - | 1 | 414 | |||
| BC104883.1 | - | 1 | 378 | 8143886 | ||
| DA041186.1 | - | 1 | 472 | |||

NAP1L5
Genomic Map