siRNA Details
Gene Symbol: CCDC88B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032251.5 | NP_115627.6 | 2 | 196 | ||
XM_006718519.3 | XP_006718582.1 | 2 | 208 | |||
XM_011544957.2 | XP_011543259.1 | 2 | 209 | |||
XM_011544959.2 | XP_011543261.1 | 2 | 209 | |||
XM_011544960.2 | XP_011543262.1 | 2 | 208 | |||
XM_017017601.1 | XP_016873090.1 | 2 | 210 | |||
GenBank mRNA | AK090436.1 | - | 2 | 191 | ||
BC140822.1 | - | 2 | 185 | 9021690 | ||
BC140824.1 | - | 2 | 185 | 9021692 | ||
BC141866.1 | - | 2 | 175 |
