siRNA Details
| Gene Symbol: CCDC88B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032251.5 | NP_115627.6 | 9 | 923 | ||
| XM_006718519.3 | XP_006718582.1 | 9 | 935 | |||
| XM_011544957.2 | XP_011543259.1 | 9 | 936 | |||
| XM_011544959.2 | XP_011543261.1 | 9 | 936 | |||
| XM_011544960.2 | XP_011543262.1 | 9 | 935 | |||
| XM_017017601.1 | XP_016873090.1 | 9 | 937 | |||
| GenBank mRNA | AK074126.1 | - | 3 | 336 | ||
| BC140822.1 | - | 9 | 912 | 9021690 | ||
| BC140824.1 | - | 9 | 912 | 9021692 | ||
| BC141866.1 | - | 9 | 902 | |||

CCDC88B
Genomic Map