siRNA Details
| Gene Symbol: DNHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144666.2 | NP_653267.2 | 9 | 2308 | ||
| NM_173589.3 | NP_775860.3 | 8 | 2411 | |||
| GenBank mRNA | AK093028.1 | - | 5 | 1492 | ||
| BC111765.1 | - | 7 | 2161 | |||
| BX648742.1 | - | 8 | 2406 | |||
| DB074960.1 | - | 2 | 270 | |||

DNHD1
Genomic Map