siRNA Details
| Gene Symbol: ZNF740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004304.3 | NP_001004304.1 | 5 | 746 | ||
| XM_006719344.2 | XP_006719407.1 | 5 | 721 | |||
| XM_011538194.2 | XP_011536496.1 | 5 | 946 | |||
| XM_017019181.1 | XP_016874670.1 | 5 | 980 | |||
| GenBank mRNA | AK091946.1 | - | 2 | 129 | ||
| AK292744.1 | - | 5 | 797 | |||
| BC053557.1 | - | 5 | 749 | 6162269 | ||

ZNF740
Genomic Map