siRNA Details
| Gene Symbol: B4GALNT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173593.3 | NP_775864.3 | 4 | 426 | ||
| GenBank mRNA | AB089940.1 | - | 4 | 426 | ||
| AK093503.1 | - | 2 | 148 | |||
| CN400443.1 | - | 4 | 333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B4GALNT3| Gene Symbol: B4GALNT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173593.3 | NP_775864.3 | 4 | 426 | ||
| GenBank mRNA | AB089940.1 | - | 4 | 426 | ||
| AK093503.1 | - | 2 | 148 | |||
| CN400443.1 | - | 4 | 333 | |||