siRNA Details
| Gene Symbol: C12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031748.3 | NP_001026918.2 | 8 | 1197 | ||
| NM_001319247.1 | NP_001306176.1 | 8 | 1197 | |||
| NR_135051.1 | 7 | 1058 | ||||
| XM_005268806.3 | XP_005268863.1 | 7 | 1007 | |||
| XM_011538231.2 | XP_011536533.1 | 7 | 1234 | |||
| GenBank mRNA | AK097445.1 | - | 7 | 1007 | ||
| BC038754.1 | - | 8 | 1194 | 5269851 | ||
| BC048120.1 | - | 8 | 1181 | 5266211 | ||
| EU668334.1 | - | 7 | 1007 | |||
| GQ472216.1 | - | 7 | 1007 | |||

C12orf40
Genomic Map