siRNA Details
| Gene Symbol: TMPRSS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182559.2 | NP_872365.1 | 5 | 1015 | ||
| GenBank mRNA | BC048112.1 | - | 5 | 1000 | 4830541 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS12| Gene Symbol: TMPRSS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182559.2 | NP_872365.1 | 5 | 1015 | ||
| GenBank mRNA | BC048112.1 | - | 5 | 1000 | 4830541 | |