siRNA Details
| Gene Symbol: CHAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164144.1 | NP_001157616.1 | 3 | 2706 | ||
| NM_001164145.1 | NP_001157617.1 | 3 | 2670 | |||
| NM_032436.2 | NP_115812.1 | 3 | 2709 | |||
| GenBank mRNA | AB058705.1 | - | 1 | 2453 | ||
| AK074894.1 | - | 1 | 2173 | |||
| AK096346.1 | - | 3 | 2686 | |||
| BC004820.1 | - | 1 | 656 | |||
| BC065237.1 | - | 3 | 2659 | 6162118 | ||

CHAMP1
Genomic Map