siRNA Details
Gene Symbol: CHAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164144.1 | NP_001157616.1 | 3 | 735 | ||
NM_001164145.1 | NP_001157617.1 | 3 | 699 | |||
NM_032436.2 | NP_115812.1 | 3 | 738 | |||
GenBank mRNA | AB058705.1 | - | 1 | 482 | ||
AK096346.1 | - | 3 | 715 | |||
BC065237.1 | - | 3 | 688 | 6162118 |
