siRNA Details
| Gene Symbol: KCNRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173605.1 | NP_775876.1 | 2 | 864 | ||
| NM_199464.2 | NP_955751.1 | 3 | 962 | |||
| GenBank mRNA | AK313401.1 | - | 3 | 813 | ||
| AY129653.1 | - | 3 | 772 | |||
| AY129654.1 | - | 2 | 682 | |||
| AY169387.1 | - | 3 | 962 | |||
| AY169388.1 | - | 2 | 864 | |||
| AY190921.1 | - | 2 | 644 | |||
| AY190922.1 | - | 3 | 742 | |||
| BC020887.1 | - | 3 | 772 | 4690662 | ||
| BC032343.1 | - | 2 | 658 | 5207702 | ||

KCNRG
Genomic Map