siRNA Details
| Gene Symbol: ZDHHC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174976.2 | NP_777636.2 | 2 | 575 | ||
| XM_006720119.3 | XP_006720182.1 | 2 | 805 | |||
| XM_011536661.2 | XP_011534963.1 | 2 | 4444 | |||
| GenBank mRNA | AK095612.1 | - | 2 | 575 | ||
| AK294840.1 | - | 2 | 574 | |||
| BC117676.2 | - | 1 | 400 | 40069732 | ||

ZDHHC22
Genomic Map