siRNA Details
| Gene Symbol: TSSK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184739.1 | NP_001171668.1 | 4 | 1177 | ||
| NM_001308067.1 | NP_001294996.1 | 4 | 872 | |||
| NM_174944.3 | NP_777604.2 | 4 | 1147 | |||
| XM_011536663.2 | XP_011534965.1 | 4 | 1029 | |||
| XM_017021226.1 | XP_016876715.1 | 4 | 1415 | |||
| GenBank mRNA | AF542390.1 | - | 2 | 1140 | ||
| AK131042.1 | - | 4 | 1177 | |||
| AY461663.1 | - | 4 | 1111 | |||
| BC111088.1 | - | 4 | 809 | 40036837 | ||
| BI825382.1 | - | 2 | 219 | |||

TSSK4
Genomic Map