siRNA Details
| Gene Symbol: TSSK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184739.1 | NP_001171668.1 | 1 | 337 | ||
| NM_174944.3 | NP_777604.2 | 1 | 337 | |||
| XM_017021226.1 | XP_016876715.1 | 1 | 533 | |||
| GenBank mRNA | AK131042.1 | - | 1 | 337 | ||
| AY461663.1 | - | 1 | 301 | |||

TSSK4
Genomic Map