siRNA Details
| Gene Symbol: FAM177A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079519.1 | NP_001072987.1 | 4 | 628 | ||
| NM_001289022.1 | NP_001275951.1 | 3 | 351 | |||
| NM_173607.3 | NP_775878.2 | 2 | 298 | |||
| GenBank mRNA | AK096173.1 | - | 3 | 239 | ||
| AL833508.1 | - | 2 | 212 | |||
| BC029559.1 | - | 3 | 220 | 5310901 | ||
| BG700755.1 | - | 3 | 351 | |||
| BX640884.1 | - | 2 | 301 | |||
| BX640984.1 | - | 2 | 249 | |||

FAM177A1
Genomic Map