siRNA Details
| Gene Symbol: FAM169B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182562.2 | NP_872368.2 | 5, 6 | 616 | ||
| GenBank mRNA | AK097062.1 | - | 5, 6 | 616 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM169B| Gene Symbol: FAM169B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182562.2 | NP_872368.2 | 5, 6 | 616 | ||
| GenBank mRNA | AK097062.1 | - | 5, 6 | 616 | ||