siRNA Details
| Gene Symbol: FBXL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203373.2 | NP_976307.2 | 1 | 220 | ||
| XM_005254320.3 | XP_005254377.1 | 1 | 1348 | |||
| XM_006720478.1 | XP_006720541.1 | 1 | 56 | |||
| XM_011521468.2 | XP_011519770.1 | 1 | 186 | |||
| XM_011521469.1 | XP_011519771.1 | 1 | 182 | |||
| XM_011521472.2 | XP_011519774.1 | 1 | 186 | |||
| XM_017022086.1 | XP_016877575.1 | 1 | 1348 | |||
| GenBank mRNA | AA495872.1 | - | 1 | 220 | ||
| AK096945.1 | - | 1 | 182 | |||
| BC065833.1 | - | 1 | 176 | 30390268 | ||

FBXL22
Genomic Map